missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(III), for analysis, ExpertQ, Scharlab

Product Code. 11394234
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394234 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394234 Supplier Scharlab Supplier No. PO02430500
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 13746-66-2
Quantity 500 g
Molecular Formula C6FeK3N6
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tripotassium hexacyanoirontris(ylium)
Molecular Weight (g/mol) 329.25
Grade ExpertQ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.