Learn More
Potassium Disulfate, EMSURE™, MilliporeSigma™,
Product Code. 15897320
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Plastic Drum
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15897320 50 kg Plastic Drum 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15897320 Supplier Supelco Supplier No. 1051079050
 Restricted Item
Estimated Shipment: 26-11-2026
Specifications
Melting Point 325°C
CAS 7790-62-7
Color White
pH 0.8 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 50 kg
Molecular Formula K2O7S2
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfonato sulfate
Molecular Weight (g/mol) 254.31
PubChem CID 62681
Formula Weight 254.33
Percent Purity 97.5 to 100.0%
Grade ACS
Packaging Plastic Drum
Chemical Name or Material Potassium Pyrosulfate