missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Supelco™

Product Code. 15854240
Change view
Click to view available options
Quantity:
80 g
Unit Size:
80g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854240 80 g 80g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15854240 Supplier Supelco Supplier No. 1024030080
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Volumetric standard, secondary reference material for redox titration, traceable to NIST SRM Certipur™ Reag. USP

Potassium dichromate is a volumetric standard manufactured under stringent conditions and measured with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point >500°C/1013 hPa
Melting Point 398°C
CAS 7778-50-9
Quantity 80 g
MDL Number MFCD00011367
Synonym Potassium dichromate, Potassium bichromate, Potassium pyrochromate, Potassium dichromate
Molecular Weight (g/mol) 294.18
Grade Research
Density 2.7 g/cm3 at 20°C
Chemical Name or Material Potassium Dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.