missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 4% (w/v), Ricca Chemical

Product Code. 31940475
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940475 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31940475 Supplier Ricca Chemical Company Supplier No. R6042800500A
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

Color Orange
CAS 7778-50-9, 7732-18-5
CAS Min % 94.0, 3.77
CAS Max % 98.0, 3.93
Molecular Formula Cr2K2O7
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 294.182
PubChem CID 24502
ChEBI CHEBI:53444
Percent Purity 4%
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼3.5 to 4
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 4% (w/v)
Chemical Name or Material Potassium Dichromate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.