Learn More
Potassium Deuteroxide (40% w/w in D2O), TRC
Product Code. 30424447
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424447 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424447 Supplier Toronto Research Chemicals Supplier No. TRCP69506210G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Potassium Deuteroxide
CAS 24572-01-8
Molecular Formula D K O
Quantity 10 g
Synonym Potassium hydroxide (K(OD)) (6CI,7CI,8CI,9CI), Potassium deuteroxide, Potassium hydroxide-d
InChI Formula InChI=1S/K.H2O/h;1H2/q+1;/p-1/i/hD
SMILES [2H]O[K]
IUPAC Name deuteriooxypotassium
Molecular Weight (g/mol) 57.112
Formula Weight 56.973
Concentration or Composition (by Analyte or Components) 40 %