Learn More
Potassium Chromate Solution, Chem Lab™
Product Code. 11988514
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11988514 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11988514 Supplier Chem-Lab Supplier No. CL02.1118.1000
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Specifications
Color Orange
Boiling Point 100°C
Melting Point 0°C
Composition 100 g K2CrO4 / l H2O
Density 1.01g/mL
Solubility Information Water Soluble
Odor Odorless
pH 8 to 10
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium chromate 10% solution
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium dichromate