missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride solution, MilliporeSigma™
Specifications
Specifications
| CAS | 7447-40-7 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Odor | Odorless |
| Packaging | Plastic Bottle |
| Density | 1.13g/cm3 at 20°C |
| Molecular Formula | ClK |
| MDL Number | MFCD00011360 |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| Solubility Information | Soluble in water at 20°C |
| Show More |
Safety and Handling
Product Identifier
- Potassium chloride solution 3 mol/l
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?