Learn More
Potassium Chloride Solution Certified Reference Material, Supelco™
Description
This certified reference material (CRM) is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany. The electrolytic conductivity of this standard is determined using a conductivity meter from an accredited Merck calibration laboratory. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search to view certified values, including uncertainty and expiry date.
Specifications
Specifications
| CAS | 7447-40-7 |
| Boiling Point | 100°C/1013 hPa |
| Molecular Formula | KCl |
| MDL Number | MFCD00011360 |
| Synonym | Potassium chloride solution, Potassium chloride solution |
| Molecular Weight (g/mol) | 74.55 |
| Quantity | 30 x 30 mL |
| Chemical Name or Material | Potassium Chloride Solution Certified Reference Material |