missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, ACS Reagent Grade, Scharlab

Product Code. 11364224
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364224 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11364224 Supplier Scharlab Supplier No. PO01671000
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in analytical chemistry, as a laboratory reagent and in photography.

  • Molecular Formula: KBr
  • Molar Mass: 119.01g/mol
  • CAS: 7758-02-3
  • EIN ECS No.: 231-830-3
  • Bromates: 0.001% max
  • Iodates (IO3): 0.001% max
  • Iodides (I): 0.001% max

Specifications

Boiling Point 1380°C
Melting Point 730°C
CAS 2-3-7758
pH 5.0–8.8 (5%, water)
Quantity 1 kg
Molecular Formula BrK
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
ChEBI CHEBI:32030
Formula Weight 119.01g/mol
Percent Purity 99.5% min (argentometric)
Grade ACS Reagent
Identification Passes Test
Loss on Drying 0.5% max (105°C)
Vapor Pressure 1.3hPa (at 795°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.