missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate, Supelco™

Product Code. 32238186
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238186 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238186 Supplier Supelco Supplier No. 1049129025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For analysis (max 0,000001% Hg) EMSURE™ ACS,ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium bromate
Melting Point 409°C to 413°C
CAS 7758-01-2
Quantity 25 kg
Density 3.13 g/cm³ at 20°C
Synonym Potassium bromate, Bromic Acid, Potassium Salt, Potassium bromate
Molecular Weight (g/mol) 167
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.