missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimony(III) Oxide Tartrate Trihydrate, Supelco™

Product Code. 15835090
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15835090 250 g 250g
15827640 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15835090 Supplier Supelco Supplier No. 1080920250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5

Specifications

Quantity 250 g
Molecular Formula K2(SbO)2C8H4O10 · 3 H2O
MDL Number MFCD00167056
Synonym Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium tartrate, Potassium antimony tartrate, Tartar emetic, Potassium antimony(III) oxide tartrate trihydrate
Molecular Weight (g/mol) 667.87
Grade Research
Density 2.6 g/cm3 at 20°C
Chemical Name or Material Potassium antimony(III) Oxide Tartrate Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.