Learn More
Brady™ Posts and Rubber Bases
Product Code. 11771306 Shop All Brady Products
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11771306 Red 1 pack
11721306 Red on White 1 pack
11731306 Black and Yellow Striped 1 pack
11751306 White 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11771306 Supplier Brady™ Supplier No. 256730
  • ONLINE EXCLUSIVE PRICE
    133.00 EUR
    122.93 EUR / Pack of 1
    Save 10.07 EUR
    7% Off
Estimated Shipment: 20-11-2026
Posts and Rubber Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Post and Fillable Base.
Specifications
Alternate Part Number POST & RUBBER BASE RED PK 4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color Red
Type Post with Rubber Base