Learn More
Brady™ Post and Cement Bases
Product Code. 11721316
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11721316 Red on White 1 pack
11791306 Red 1 pack
11711316 Black and Yellow Striped 1 pack
11741316 White 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11721316 Supplier Brady™ Supplier No. 292237
  • ONLINE EXCLUSIVE PRICE
    164.00 EUR
    151.89 EUR / Pack of 1
    Save 12.11 EUR
    7% Off
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
These Post and Cement Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Includes Post and Cement Base.
Specifications
Alternate Part Number BRADYPOST & CEMENT BASE RED/WHT PK4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color Red on White
Type Post with Cement Base