Learn More
Globe Scientific PosiStop LTS-Compatible Pipette Tips, Empty Racks
Shop All Globe Scientific ProductsDescription
- Empty racks for 20 μL, 200 μL, and 1000 μL PosiStop pipette tips
- Color-coded trays: red (20 μL), green (200 μL), blue (1000 μL)
- Durable and reusable for efficient lab workflows
- Ideal for refilling and organized tip storage
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Natural |
| Holds | Pipette Tips (1000 μL) |
| Material | Polypropylene |
| Positions | 96 |
| Shape | Rectangular |
| Stackable | Yes |
| Sterility | Non-sterile |
| Product Type | Empty Rack |
| Array | 12 x 8 |