Learn More
Thermo Scientific Chemicals Posaconazole
Product Code. 17201226 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 17201226
Brand: Thermo Scientific Chemicals 467660050
Request Bulk or Custom Format
CAS: 171228-49-2 | C37H42F2N8O4, C37H42F2N8O4 | 700.79 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 171228-49-2
Molecular Formula C37H42F2N8O4
Specifications
Quantity 5 g
Melting Point 170.0°C to 179.0°C
Formula Weight 700.79
InChI Key RAGOYPUPXAKGKH-XAKZXMRKSA-N
SMILES CC[C@@H]([C@H](C)O)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=NC=N3)(C2)C2=CC=C(F)C=C2F)C=C1
IUPAC Name 4-{4-[4-(4-{[(3R,5R)-5-(2,4-difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-1-[(2S,3S)-2-hydroxypentan-3-yl]-4,5-dihydro-1H-1,2,4-triazol-5-one
Molecular Weight (g/mol) 700.79
Chemical Name or Material Posaconazole
compliance-icons
Product Identifier
  • Posaconazole
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Posaconazole