Learn More
OHAUS™ Portable Balance - CR Series
Product Code. 16606772 Shop All Ohaus Products
Change view
Click to view available options
Model:
CR2200
CR221
CR5200
CR621
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
16606772 CR5200 Each
16615164 CR221 Each
16625164 CR621 Each
16635164 CR2200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16606772 Supplier OHAUS™ Supplier No. 30428216
Add to basket
Add to basket
Quality Portable Electronic Scales Suitable for Everyday Weighing.
  • Intuitive two-button design makes for easy operation.
  • Slim, space-saving design allows for stackable storage.
  • Enlarged weighing platform can adequately handle large samples and a variety of objects.
TRUSTED_SUSTAINABILITY
Specifications
Description Stability indicator, mechanical and software overload/underload protection, auto shut-off
Capacity 5200.00 g
Display Type LCD
Readability 1.0000 g
For Use With (Application) Weighing
Model CR5200
Stabilization Time 2.000 s
Weighing Modes Gram, Pound, Ounce, Ounce Troy, & Pennyweight
Dimensions (L x W x H) 23.500 x 17.000 x 5.500 cm
Length (English) 8.10 in.
Length (Metric) 205.00 mm
Width (English) 5.50 in.
Width (English) Pan 5.000 in.
Width (Metric) 140.00 mm
Width (Metric) Pan 125.000 mm
Height (English) 1.60 in.
Height (Metric) 41.000 mm
Depth (English) 8.10 in.
Depth (English) Pan 5.200 in.
Depth (Metric) 205.00 mm
Depth (Metric) Pan 132.000 mm
Weight (English) 0.900 lb.
Weight (Metric) 0.400 kg
Electrical Requirements 3 AA batteries (included) or AC adapter (not included)