Learn More
Haldenwanger™ Porcelain Buchner Funnel
Product Code. 10623491
Change view
Click to view available options
Capacity (Metric):
10 mL
10,600 mL
1250 mL
25 mL
3 mL
4300 mL
5800 mL
Diameter (Metric) Plate:
19 mm
29 mm
42 mm
155 mm
250 mm
279 mm
330 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Plate Unit Size
10623491 10 mL 29 mm Each
10303081 3 mL 19 mm Each
10781922 25 mL 42 mm Each
10027440 1250 mL 155 mm Each
10695021 4300 mL 250 mm Each
10642911 5800 mL 279 mm Each
10579021 10,600 mL 330 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10623491 Supplier Haldenwanger™ Supplier No. L 127C/00
  • ONLINE EXCLUSIVE PRICE
    46.70 EUR
    43.53 EUR / Pack of 1
    Save 3.17 EUR
    6% Off
Estimated Shipment: 09-12-2026
Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Available with perforated fixed filter plate
  • Complies with DIN VDE 0335, type C 100
Specifications
Capacity (Metric) 10 mL
Material Porcelain
Height (Metric) 60 mm
Diameter (Metric) Plate 29 mm
Diameter (Metric) Disc 29 mm
Quantity 1Each
Product Type Buchner Funnel