missing translation for 'onlineSavingsMsg'
Learn More

Ponazuril, TRC

Product Code. 17933145
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933145 100 mg 100mg
17904705 10 mg 10mg
17923145 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933145 Supplier Toronto Research Chemicals Supplier No. TRCP689425100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69004-04-2
Molecular Formula C18 H14 F3 N3 O6 S
Molecular Weight (g/mol) 457.38
Synonym Ponazuril, 1-Methyl-3-[3-methyl-4-[4-[(trifluoromethyl)sulfonyl]phenoxy]phenyl]-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, BAY-Vi 9143, Ponazuril, Toltrazuril sulfone
IUPAC Name 1-methyl-3-[3-methyl-4-[4-(trifluoromethylsulfonyl)phenoxy]phenyl]-1,3,5-triazinane-2,4,6-trione
SMILES CN1C(=O)NC(=O)N(C1=O)c2ccc(Oc3ccc(cc3)S(=O)(=O)C(F)(F)F)c(C)c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H14F3N3O6S/c1-10-9-11(24-16(26)22-15(25)23(2)17(24)27)3-8-14(10)30-12-4-6-13(7-5-12)31(28,29)18(19,20)21/h3-9H,1-2H3,(H,22,25,26)
Formula Weight 457.0555
Percent Purity >95%
Chemical Name or Material Ponazuril
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.