Learn More
MedChemExpress Pomalidomide-PEG4-C-COOH
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 507.49 g/mol |
| CAS | 2097938-44-6 |
| Content And Storage | 4°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen) |
| Shipping Condition | Room temperature |
| Quantity | 1 g |
| Synonym | Cereblon Ligand -Linker Conjugates 1 ∣ E3 Ligase Ligand-Linker Conjugates 1 |
| Solubility | DMSO : 50 mg/mL (98.52 mM; Need ultrasonic) ∣ Ethanol : 25 mg/mL (49.26 mM; Need ultrasonic) |
| Target | HBV |