missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinyl Acetate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 9003-20-7 |
| Molecular Formula | (C4H6O2)n |
| MDL Number | MFCD00084457 |
| Synonym | vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer |
| Quantity | 100 g |
| InChI Key | XTXRWKRVRITETP-UHFFFAOYSA-N |
| SMILES | CC(=O)OC(-*)C-* |
| IUPAC Name | ethenyl acetate |
| Molecular Weight (g/mol) | 86.09 |
| PubChem CID | 7904 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction