Learn More
Nitritex™ Polyurethane Tacky Mat
Product Code. 15268318
Change view
Click to view available options
Color:
Blue
White
Dimensions (L x W):
18 x 45 in.
26 x 45 in.
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Unit Size
15268318 White 26 x 45 in. Pack of 4
15258318 White 18 x 45 in. Pack of 4
15238318 Blue 26 x 45 in. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15268318 Supplier Nitritex™ Supplier No. STW2645
  • ONLINE EXCLUSIVE PRICE
    199.50 EUR
    122.59 EUR / Pack of 4
    Save 76.91 EUR
    38% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Keep dirt, dust and bacteria at bay in controlled laboratory environments with the Nitritex™ Polyurethane Tacky Mat.
  • Matting for use in controlled laboratory environments
  • Rectangular
  • Polyurethane
  • White or blue
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001:2015
Dimensions (L x W) 26 x 45 in.
Quantity 30 Sheets/Mat, 4 Mats/Cs.
Color White
Material Polyurethane