Learn More
mb2 Polyurethane Perforated Chair on Gliders - Asynchronus Mecanism
Product Code. 30130639
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30130639 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30130639 Supplier mb2 Supplier No. AP3PEXB1D1E3
  • ONLINE EXCLUSIVE PRICE
    342.00 EUR
    325.31 EUR / Pack of 1
    Save 16.69 EUR
    4% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Polyurethane perforated chair on gliders - Normal lift - Asynchronus mechanism
  • Chair in perforated polyurethane
  • Seat and back in perforated polyurethane
  • Asynchronus mecanism, adjustable seat and backrest
  • Pneumatic cylinder low
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Gliders
Specifications
Product Type Laboratory Seat
Color Black
Adjustable (Height) Yes
Autoclavable Not Autoclavable
Back Style Standard
Quantity 1 Ea.
Seating Height 390 to 510 mm
Weight (Metric) 14.44 kg
With Armrest No
With Backrest Yes
With Casters No
Footrest No
With Lumbar Support No
With Tilt Yes
Material Polyurethane
Seating Type Desk Height