Learn More
Fisherbrand™ True North™ Polyurethane Foam Ice Bucket
Product Code. 11314085 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11314085 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11314085
  • ONLINE EXCLUSIVE PRICE
    70.20 EUR
    61.03 EUR / Pack of 1
    Save 9.17 EUR
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Keep your temperature-sensitive reagents and samples cool while working at the bench. True North™ Polyurethane Foam Ice Bucket has a snug-fitting lid to help reduce evaporation and nests underneath the container when not in use.
  • Durable and light-weight black polyurethane foam with metal inserts.
  • Snug-fitting lid helps to reduce evaporation and spilling.
  • Nests underneath the container when not in use to help prevent loss.
  • Convenient spout helps prevent spills.
Specifications
Height (Metric) 181 mm
Height (English) 7.1 in.
Length (Metric) 234 mm
Length (English) 9.2 in.
Closure Type Lid
Capacity (English) 1.1 gal.
For Use With (Equipment) Dry ice, ice water, ice-salt slurries, etc.
Includes Lid, Spout, Side Carrying Handles
Material Polyurethane Foam
Product Type Ice Bucket
Quantity 1 Ea.
Color Black
Capacity (Metric) 4 L
Holds Ice (4 L)
Autoclavable Not Autoclavable
Temperature Control No
Temperature Range -80°C to +240°C