Learn More
Fisherbrand™ Polyurethane Foam Stoppers
Product Code. 12974291 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
15 mm
25 mm
30 mm
35 mm
40 mm
55 mm
70 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12974291 35 mm Pack of 100
12944311 15 mm Pack of 100
12954311 25 mm Pack of 100
12964291 30 mm Pack of 100
12984291 40 mm Pack of 100
12904301 55 mm Pack of 100
12914301 70 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12974291
  • ONLINE EXCLUSIVE PRICE
    17.85 EUR
    12.06 EUR / Pack of 100
    Save 5.79 EUR
    32% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Formed by polymerization in the presence of a foaming agent.
  • Made up of polyurethane
  • Rigid and flexible
Specifications
Diameter (Metric) 35 mm
Height (Metric) 50 mm
Quantity 100/Pk.
Material Polyurethane
Product Type Plug Stopper