Learn More
Cytiva Polyethylene (PE) Capillary Tubing
Product Code. 11320022 Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
1.1 mm
Unit Size:
Each
Product Code. 11320022
Brand: Cytiva 19004101
Only null left
Add to basket
Add to basket
For use with chromatography applications
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 1.1 mm
Material Polyethylene
Diameter (Metric) Outer 1.8 mm
Overall Length (Metric) 5000 mm
Packaging Quantity 1 Pack