Learn More
VITLAB™ Polystyrene Non-sterile Pipets
Product Code. 16563592
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
Unit Size:
Pack of 100
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16563592 10 mL Pack of 100
16503592 1 mL Pack of 200
16523592 2 mL Pack of 200
16543592 5 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16563592 Supplier VITLAB™ Supplier No. 161019
Estimated Shipment: 11-11-2026
Specifications
Material Polystyrene
Graduated Yes
Graduations 0.10 mL
Quantity 10 Pack
Color Transparent
Volume (Metric) 10 mL
Type Pipet