Learn More
VITLAB™ Polystyrene Non-sterile Pipets
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Non-sterile |
| Graduated | Yes |
| Graduations | 0.01 mL |
| Quantity | 10 Pack |
| Color | Transparent |
| Volume (Metric) | 1 mL |
| Length (Metric) | 272 mm |
| Type | Pipet |