Learn More
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Product Code. 10349334 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
4 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10349334 4 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10349334
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
Cuvets with improved photometric properties.
  • Very low variation of extinction values
  • Excellent optical transmission range
Specifications
Type Macro Cuvette
Disposable Reusable
Capacity (Metric) 4 mL
Material Plastic
Quantity 100/Pk.
Wavelength Range 220 to 900 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm