Learn More
Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water
Description
These particles are widely used in phagocytosis and neural retrograde transport studies, and as markers for cell bound antigens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 9003-53-6 |
| Physical Form | Dispersion |
| Quantity | 15 mL |
| Odor | Odorless |
| Molecular Formula | (C8H8)n |
| MDL Number | MFCD00084450 |
| Synonym | ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene |
| Solubility Information | Fully miscible in water. |
| InChI Key | PPBRXRYQALVLMV-UHFFFAOYSA-N |
| SMILES | *-CC(-*)C1=CC=CC=C1 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.