Learn More
Corning™ Polystyrene Adapter Applicator
Product Code. 10062521 Shop All Corning Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10062521 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10062521 Supplier Corning™ Supplier No. 3507
  • ONLINE EXCLUSIVE PRICE
    3320.00 EUR
    3102.30 EUR / Pack of 1
    Save 217.70 EUR
    6% Off
Estimated Shipment: 03-03-2027
Applications include: PCR clean-up; dye-terminator clean-up and separation/filtration assays
With a well volume of 360μL the plate body is made of white polystyrene with a K-Resin™ insert. The standard membrane consists of a low protein binding, hydrophilic 0.2μm PVDF (Polyvinylidene Fluoride). Another feature is the individual filter discs. These are encapsulated by a special over-moulding process that ensures the integrity of each well and eliminates liquid cross contamination, while the optimised nozzle design prevents cross-contamination of the filtrate. The plate has the standard industry (SBS) footprint, so is compatible with most automation, liquid handling and detection equipment. Main applications include: PCR* clean-up; dye-terminator clean-up and separation/filtration assays.
TRUSTED_SUSTAINABILITY
Specifications
Membrane Hydrophilic 0.2μm Polyvinylidene Fluoride
Array 8 x 12
Color White
For Use With (Equipment) PCR clean-up; dye-terminator clean-up and separation/filtration assays
Material Polystyrene
No. of Wells 96
Total Volume (Metric) Well 360 μL
For Use With (Application) PCR clean-up, dye-terminator clean-up and separation/filtration assays
Quantity Each