Learn More
Greiner Bio-One Polystyrene 96-well Half Area Microplates
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Traceable to production through a defined LOT number system
- μClear™ bottom
Specifications
Specifications
| Certifications/Compliance | DIN ISO 9001 |
| Certified | Yes |
| Format | Half-Area |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Show More |