Learn More
Kartell™ Plastilab™ Unequal Straight Tapered Tubing Connectors
BIOPHARMA
Product Code. 11924289
Change view
Click to view available options
Port Size:
1.6/7.5mm
Bore Size: 1.6 to 4.6mm
Bore Size: 1.6 to 7.5mm
Bore Size: 4.6 to 7.5mm
Unit Size:
Pack of 10
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Port Size Unit Size
11924289 Bore Size: 1.6 to 7.5mm Pack of 10
11914289 Bore Size: 1.6 to 4.6mm Pack of 10
11934289 Bore Size: 4.6 to 7.5mm Pack of 10
12513317 1.6/7.5mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11924289 Supplier Kartell™ Supplier No. 7087800
  • ONLINE EXCLUSIVE PRICE
    21.00 EUR
    12.54 EUR / Pack of 10
    Save 8.46 EUR
    40% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Designed with different inlet and outlet diameters.
  • For use with flexible tubing
  • Autoclavable
Specifications
Diameter (Metric) 4-8 mm to 12-16 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With For flexible tube
Packaging Quantity 10/Pk.
Port Size Bore Size: 1.6 to 7.5mm
Shape Straight
Product Type Unequal Straight Tapered Tubing Connector