Learn More
Multiroir™ Block of Polystyrene Trays (6), 32102PTGMB
Product Code. 11778371
Change view
Click to view available options
Capacity (Metric):
80 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11778371 80 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11778371 Supplier Multiroir™ Supplier No. 32102
  • ONLINE EXCLUSIVE PRICE
    117.00 EUR
    115.02 EUR / Pack of 1
    Save 1.98 EUR
    1% Off
Estimated Shipment: 19-10-2026
Multi-trayed block of six variegated color trays/drawers suit a variety of sorted storage needs. Multiroir™ Block of Polypropylene Trays (6), 32102PTGMB can be equippped with a variety of thermoformed honeycomb grids.
  • Polystyrene construction
  • Variegated colors - one color for each tray/drawer
  • Transportable
  • Stackable
  • Variety of honeycomb grids for drawers available separately
Order Info
CapacityMetric
Specifications
Capacity (Metric) 80 L
Description 340 x 250 x 40mm (internal per drawer)
Color Beige, Blue, Gray, Green, Red, White, Yellow
Material Polystyrene
Shape Rectangular
Mounting Type Stackable
No. of Compartments 6 Trays/Drawers
Product Type Storage Box
Quantity 1/Ea.
Stackable Yes
Dimensions (L x W x H) 365 x 266 x 310 mm (external)
Length (Metric) 365 mm
Width (Metric) 266 mm
Height (Metric) 310 mm