Learn More
Fisherbrand™ Polypropylene Transparency Makro Pipet Tips
Product Code. 11846943 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1000 to 5000 μL
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11846943 1000 to 5000 μL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11846943
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
Use these pipet tips with Gilson™ Pipetman™/Neo Pipets. Fisherbrand™ Polypropylene Transparency Makro Pipet Tips feature uniform high quality and precision, a perfect fit for manual or electronic, single or multichannel pipets.
Designed for use when transferring large sample volume. Advanced moulding technology ensures tips are straight. Alternative option to serological pipettes. Non-sterile, certified RNase, DNase, DNA and pyrogen free.
Specifications
Volume (Metric) 1000 to 5000 μL
Sterility Non-sterile
Color Natural
For Use With (Application) For Use When Transferring Large Sample Volume
Product Line Makro