Learn More
Ark Plas Products™ Polypropylene Threaded Male Luer
Product Code. 11771976
Change view
Click to view available options
Thread Size:
1/4-28 UNF
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
11771976 1/4-28 UNF Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11771976 Supplier Ark Plas Products™ Supplier No. LGX86PP0
  • ONLINE EXCLUSIVE PRICE
    91.08 EUR
    5.49 EUR / Pack of 25
    Save 85.59 EUR
    93% Off
View all promotions
Estimated Shipment: 04-01-2027
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Polypropylene Threaded Male Luers feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
  • Natural polypropylene luers
  • Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • Molded in virgin, lot-traceable material
  • Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Autoclavable Autoclavable
For Use With Flexible tubing or threaded ports
Min. Operating Temperature (Metric) -17°C
Packaging Quantity 25/Pk.
Port Size 1/4-28 UNF
Thread Size 1/4-28 UNF
Color Natural
Max. Operating Temperature (Metric) 135°C