Learn More
Kartell™ Polypropylene Test Tubes
Product Code. 12004598
Change view
Click to view available options
Capacity (Metric):
15 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12004598 15 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12004598 Supplier Kartell™ Supplier No. 8830304
  • ONLINE EXCLUSIVE PRICE
    134.00 EUR
    125.71 EUR / Pack of 1000
    Save 8.29 EUR
    6% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Supplied with fitted screw cap designed to avoid contamination resulting from drops of liquid on the inner surface. Kartell™ Polypropylene Test Tubes meet test tube needs and are suitable for centrifugation.
  • Polyethylene quick closure screw cap for leak-proof containment
  • Screw caps available in a variety of colors for classification
  • Dimensions: 16 dia. x 100mmH
  • CE marked
  • IVD approved
Specifications
Certifications/Compliance CE–IVD
Description 4,000 x g
Capacity (Metric) 15 mL
Diameter (Metric) 16 mm