Description
- Manufactured from polypropylene
- Includes tip tray
- Empty, for filter and pipet tips
Specifications
Specifications
| Color | Green |
| Holds | Pipette Tips (300 μL) |
| Material | Polypropylene |
| Stackable | Yes |
| For Use With (Equipment) | Pipet and Filter Tips |
| No. per Pack | 1 Pack |
| Quantity | 1 Pack |
| Product Type | Empty Tip Box |