Learn More
Chemsplash Polypropylene Socks (Pair) Elastic at Opening
Description
40GSM Polypropylene Boot
- 40GSM Polypropylene Material
- Elastic at Opening
- 36 x 18 cm
- 10 pairs/pack
- PFAS Free
Description: This product is designed for use in BRC approved work areas and the pharmaceutical industry. Made from 40GSM polypropylene material, it features an elasticated opening for a secure fit. The size is 36 x 18 cm with 10 pairs per pack. This boot is PFAS free and ensures safety and compliance with industry standards.
Specifications
Specifications
| Size | Universal |
| Material | Polypropylene |
| Latex Free | Yes |
| Coating | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Plain |
| Dimensions (L x W) | 36 x 18 cm |
| Show More |