Learn More
Macherey-Nagel™ Polypropylene Screw Caps with Hole
Product Code. 15914880 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15914880 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15914880 Supplier Macherey-Nagel™ Supplier No. 702113
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
In Stock
Polypropylene screw closures for sample storage with an analytically pure Silicone/PTFE septum that is firmly connected to the cap.
  • Closed top
  • White
  • Whie silicone/blue PTFE pre-fitted septa
Specifications
Description Screw Caps with Silicone PTFE Septa
Color White
Material Polypropylene
Septum White Silicone/Blue PTFE
For Use With (Application) Sample Storage
Temperature Range (Metric) Temp. Resistance Septa -60°C up to 200°C - However, PP cap only up to 120°C/Analytical Purity - High
Thickness (Metric) 1.3 mm (Septa)
Product Type Screw Cap