Learn More
VITLAB™ Polypropylene Reagent Bottles
Product Code. 15261159
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15261159 250 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15261159 Supplier VITLAB™ Supplier No. 100489
  • ONLINE EXCLUSIVE PRICE
    68.15 EUR
    57.72 EUR / Pack of 20
    Save 10.43 EUR
    15% Off
Estimated Shipment: 23-11-2026
Specifications
Capacity (Metric) 250 mL
Material Polypropylene
Mouth Narrow
Closure Type Screw Cap
Product Type Bottle