Learn More
Polypropylene Ratchet Clips
Product Code. 12376347
Change view
Click to view available options
Packaging Quantity:
10 Pack
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
12376347 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12376347 Supplier S Murray SAMCO Supplier No. L140/02
  • ONLINE EXCLUSIVE PRICE
    3.55 EUR
    2.88 EUR / Pack of 10
    Save 0.67 EUR
    18% Off
View all promotions
Estimated Shipment: 12-01-2027
Use these clips for a variety of laboratory applications. S Murray™ Polypropylene Ratchet Clips are suitable for use with flexible tubing up to 10mm diameter.
  • Polypropylene
  • Pack of 10
Specifications
Material Polypropylene
For Use With Suitable for use with flexible tubing up to 10mm dia.
For Use With (Application) Suitable for use with flexible tubing up to 10mm diameter
Packaging Quantity 10 Pack
Product Type Ratchet Clip