Learn More
Eppendorf™ twin.tec™ PCR Plates
Product Code. 10100803 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Clear
Red
Skirt Style:
Semi
Skirted
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Unit Size
10100803 Red Skirted Pack of 25
10767294 Blue Semi Pack of 25
10355672 Blue Skirted Pack of 25
10690744 Clear Semi Pack of 25
10120092 Clear Skirted Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10100803 Supplier Eppendorf™ Supplier No. 0030128680
In Stock
Add to basket
Add to basket
Bulk Quote Request
Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Equipment) Real time PCR
Skirt Style Skirted
Material Polypropylene, Polycarbonate
No. of Wells 96
Sterility Sterile
Total Volume (Metric) Well 150 μL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Ideal for quantitative real time PCR and fit most thermal cyclers
Quantity 25/Pk.
RNase Free Yes