Learn More
Fisherbrand™ Polypropylene Microcentrifuge Tube
Product Code. 11532293
Change view
Click to view available options
Capacity (Metric):
1.5 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11532293 1.5 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11532293 Supplier Fisherbrand™ Supplier No. 200400P
Add to basket
Add to basket
Use these tubes for a variety of applications. Fisherbrand™ Polypropylene Microcentrifuge Tubes feature a snap top cap and frosted writing area.
  • Graduated polypropylene microcentrifuge tubes
  • Can be centrifuged up to 12,500xg
  • 1.5mL capacity
  • Pack of 500
Specifications
Cap Style Snap Cap
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Material Polypropylene
No. per Pack 500/Pk.
Volume (Metric) 1.5mL
Sterility Non-sterile
Closure Type Snap Cap