Learn More
Buerkle™ Polypropylene Measuring Scoop
Product Code. 11981491 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Length (Metric):
100 mm
135 mm
160 mm
200 mm
260 mm
315 mm
385 mm
Width (Metric):
34 mm
44 mm
55 mm
70 mm
90 mm
118 mm
134 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Width (Metric) unitSize
11981491 10 mL 100 mm 34 mm Each
10352243 25 mL 135 mm 44 mm Each
10496441 50 mL 160 mm 55 mm Each
10350782 100 mL 200 mm 70 mm Each
10448521 250 mL 260 mm 90 mm Each
10524132 500 mL 315 mm 118 mm Each
10648052 1000 mL 385 mm 134 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11981491 Supplier Buerkle™ Supplier No. 96140010
  • ONLINE EXCLUSIVE PRICE
    0.70 EUR
    0.66 EUR / Pack of 1
    Save 0.04 EUR
    5% Off
View all promotions
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Scoops and measures powders and granules. Buerkle™ Polypropylene Measuring Scoops are calibrated to accurately assess volume at the brim.
Universal graduated shovel for filling and measuring powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • White polypropylene construction
  • High chemical-resistance
  • Calibrated volume when full to the brim
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Capacity (English) 0.34 oz.
Capacity (Metric) 10 mL
Color White
Product Type Measuring Scoop
For Use With (Application) For Filling and Measuring of Powders and Granules
Width (Metric) 34 mm
Material Polypropylene
Length (Metric) 100 mm