Learn More
Brady™ Polypropylene Extra Cleats for 480/600V Lockout
Description
- Assures positive restraint for breakers with long, sliding switch throws
- Polypropylene
Specifications
Specifications
| Product Type | Extra Cleats |
| Alternate Part Number | LOCKOUT CLEAT 480/600 VOLT GENERAL USE |
| Color | Red |
| Description | Extra Cleats for 480/600V Lockout |
| For Use With (Equipment) | For 480/600V Lockout |
| Quantity | 6 Pk. |