Learn More
Buerkle™ Discharge Tube for Buerkle™ AccuOne/EnergyOne
Product Code. 11941641 Shop All Bürkle Products
Change view
Click to view available options
Pump Tube Length:
200 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pump Tube Length Unit Size
11941641 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11941641 Supplier Buerkle™ Supplier No. 59003040
Estimated Shipment: 16-12-2026
Discharge tube PP for AccuOne/EnergyOne barrel pumps.
  • Discharge tube PP for AccuOne/EnergyOne barrel pumps
TRUSTED_SUSTAINABILITY
Specifications
Opening Size (English) 0.63 in.
Opening Size (Metric) 16 mm
Pump Tube Length 200 mm
Material Polypropylene
Product Type Discharge Tube