Learn More
Vitlab™ Polypropylene Bowl
Product Code. 10232222
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
4 L
7 L
Unit Size:
Each
Pack of 3
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10232222 1 L Pack of 5
10528341 2 L Each
10321742 2 L Pack of 5
10438502 1 L Each
10351022 7 L Pack of 3
10098680 4 L Each
10496871 4 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10232222 Supplier Vitlab™ Supplier No. 42594
  • ONLINE EXCLUSIVE PRICE
    13.00 EUR
    10.69 EUR / Pack of 5
    Save 2.31 EUR
    17% Off
Estimated Shipment: 28-12-2026
Use these bowls to help prevent splashing on the work surface while working with strong reactions in uncovered vessels. VITLAB™ Polypropylene Bowls feature a broad, stable edge and circumferential standing ring on the bottom.
  • White round bowl
  • Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1 L
Material Polypropylene
Height (Metric) 70 mm
Diameter (Metric) 160 mm