Learn More
BRAND™ Urine Beaker, PP, with Screw Cap PE, CE-IVD
Product Code. 10583872 Shop All Brand Products
Change view
Click to view available options
Closure Color:
Green
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 240
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Sterility Unit Size
10583872 Yellow Sterile Pack of 240
10218361 Green Non-sterile Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10583872 Supplier BRAND™ Supplier No. 758910
  • ONLINE EXCLUSIVE PRICE
    427.00 EUR
    138.96 EUR / Pack of 240
    Save 288.04 EUR
    67% Off
View all promotions
Estimated Shipment: 17-12-2026
Manufactured from polypropylene with polyethylene cap. BRAND™ Urine Beakers with Scew Caps are made specifically for hygienic urine collection and containment. Sterile or non-sterile versions available. CE-marked according to guideline EU IVDR 2017/746.
  • PP, screw cap PE. 
  • For hygienic urine sample handling. 
  • CE marked according to guideline EU IVDR 2017/746
TRUSTED_SUSTAINABILITY
Specifications
Product Type Urine Beaker
Closure Color Yellow
Closure Type Screw Cap
Graduated Yes
Material Polypropylene
Sterility Sterile
Closure Material Polyethylene
Color Transparent
For Use With (Application) Hygienic urine sample handling
Height (Metric) 75 mm
No. per Pack 240/Pk.
Volume (Metric) 100 mL