Learn More
Chemsplash Polyprop Dust Suit Coverall, Zip Front White
Description
Polypropylene Dust Suit Coverall
- 30GSM Non-Woven Polypropylene
- Zip Front
- Elasticated Wrist/Ankle
- Hood
- Suitable for Hygiene Rules / Food Processing
- Clean Areas / Disposal Operations
- PFAS Free
Description: The Polypropylene Dust Suit Coverall is made from a 30GSM non-woven material and features a full zip front closure, elasticated wrist and ankle for a secure fit, and a hood. Individually packed for hygiene purposes, it is suitable for Hygiene Rules, Food Processing, Clean Areas, and Disposal Operations.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Small |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |