missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyoxyl 40 Stearate, TRC
Specifications
Specifications
| Chemical Name or Material | Polyoxyl 6 Stearate |
| CAS | 9004-99-3 |
| Molecular Formula | (C2H4O)nC18H36O2 |
| Quantity | 1 g |
| Synonym | 40S, 40S (polyether), 60S, 60S (polyether), Akyporox S 100, Alkasurf S 65-8, Arosurf 1855E40, Atlas G 214, Empilan CQ 100, Emulgen 3199, Estol 3723, Ethofat 60/15, Ethofat 60/20, Ethofat 60/25, Ethoxylated Stearic Acid, Eumulgin ST 8, Polystate, Polystate B, Prodhybase 4000, Prodhybase P, Ritox 52, Rokacet S 10, Rokacet S 17, Rokacet S 2, Rokacet S 24, Rokacet S 8, Ropol 24, Trydet SA 40, Unipeg 200MS, Varonic 1000MS, Varonic 1800MS, Verox S 12, Verox S 16, Verox S 18, Witconol 2711, X 489R, YMS 2 |
| InChI Formula | InChI=1S/C20H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h21H,2-19H2,1H3 |
| SMILES | CCCCCCCCCCCCCCCCCC(OCCO)=O |
| Molecular Weight (g/mol) | 328.53 |
| Formula Weight | 328.2977 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?